Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio Premium Fetal Bovine Serum
SDP

Catalog No. 11852142 Shop All Gemini Bio Products
Change view
Click to view available options
Grade:
Premium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
11-852-142 Premium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-852-142 Supplier GeminiBio Supplier No. S11150
Get 1 Extra Serum When You Buy 5
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serum that balances quality and value for general cell culture and specialized research.

  • A trusted balance of quality and value for general cell culture and specialized research
  • Low endotoxin and hemoglobin for robust performance across a broad range of cell types
  • US-origin or USDA-approved material, traceable to date and location of collection
  • Triple 0.1 μm sterile-filtered
  • Every lot sterility-, virus-, and performance-tested with a full biochemical profile
  • High lot-to-lot consistency for reproducible experiments
  • Ships on dry ice; store frozen at -20°C to -10°C
  • Formerly sold under the Atlanta Biologicals and Bio-Techne/R&D Systems brands
  • Complimentary heat inactivation upon request

Specifications

Additional Assays Biochemical profile
Certifications/Compliance ISO 13485:2016 certified quality system
Content And Storage 500 mL bottle, store frozen at -20°C to -10°C
Species Bovine
Age Fetal
Country of Origin Refer to CoA for specific origin
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Fetal bovine blood
Grade Premium
Processing Triple 0.1 μm sterile-filtered
Tested For Mycoplasma, Sterility (bacteria and fungi), Adventitious viruses per 9 CFR 113.53 (BVDV, IBRV, PI-3V, BTV, BRSV, BPV, BAV-3, BAV-5, Reovirus, Rabies)
Endotoxin Concentration ≤50.0 EU/mL
Formulation Liquid, frozen
For Use With (Application) Cell Culture
Heat Inactivated No
Hemoglobin Concentration ≤25 mg/dL
Lyophilized No
Product Line Premium
Mycoplasma Detected No
Osmolality 280 to 335 mOsm/kg
pH 6.8 to 7.8
Shipping Condition Dry Ice
Test Methods LAL assay (endotoxin); USP methods (sterility); 9 CFR 113.53 (virus)
USDA Safety Testing Included Yes
Regulatory Status RUO
Gamma-irradiated No
Show More Show Less

For cell culture, further manufacturing or research use only. Not for direct therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.