Learn More
Unibor™ Premium Gel Hand Sanitizer
Description
- Regulary tested for effectiveness against pathogens
- Produced with green tea extract and other moisturizing ingredients
- Fully registered with the US FDA, Korean KFDA, EU, Health Canada, and many more
- Free of harmful ingredients - tested annually by independent labs
- Available in small flip cap bottles and 3 pump bottle sizes
Specifications
Specifications
| Product Type | Hand Sanitizer |
| Color | Clear |
| Form | Gel |
| Certifications/Compliance | FDA Registered OTC Monograph, Part333A |
| For Use With (Application) | Eliminating Germs on Hands |
| Packaging | Pump Top Bottle |
| Volume (English) | 1 gal. |
| Volume (Metric) | 3.79 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.