Learn More
Neuromics, Inc Heat Inactivated Premium Imported Fetal Bovine Serum (USDA Approved), 500 ml is ideal for the most sensitive cells lines and can be used to culture all cell types. It is low in endotoxins.
Catalog No. NC2071920
Encompass
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance.Our Heat Inactivated Premium Imported (USDA Approved) FBS features: Endotoxin levels : ≤10 EU/mL, Triple 0.1 μm Filtered, and Free of Virus and Mycoplasma.This treatment allows to destroy the complement acting in the immune system to avoid an interference with some experimentation, in particular the tests of immunology. Excellent for research involving immune response, viral studies, cytotoxicity assays, and stem cells.
Catalog No. NC2071920 Supplier Neuromics, Inc Supplier No. FBS002HI
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance.Our Heat Inactivated Premium Imported (USDA Approved) FBS features: Endotoxin levels : ≤10 EU/mL, Triple 0.1 μm Filtered, and Free of Virus and Mycoplasma.This treatment allows to destroy the complement acting in the immune system to avoid an interference with some experimentation, in particular the tests of immunology. Excellent for research involving immune response, viral studies, cytotoxicity assays, and stem cells.