Learn More
Chemglass Life Sciences Premium Micro Homogenizer Package
SDP
Catalog No. 501311380
Encompass
Premium Micro Homogenizer Package. These micro-homogenizing packages offer the capability of processing all types of samples within micro-tubes. Whether gently mixing, extracting RNA/DNA or destroying the cell wall, these packages can provide fast and complete effective results. Sample volumes as small as 0.03mL can be easily processed in a matter of seconds and cleanup is just as easy. Please contact us for 220 volt homogenizing packages.CLS-5034-16 Premium Micro Package Contains:1 each CLS-5030-10 Homogenizer1 each CLS-5032-10 5 x 75mm Generator1 each CLS-5032-11 7 x 75mm Generator1 each CLS-5030-30 Stand Assembly
Catalog No. 50-131-1380 Supplier Chemglass Life Sciences Supplier No. CLS503416
Add to Cart
Add to Cart
Request a Quote
Premium Micro Homogenizer Package. These micro-homogenizing packages offer the capability of processing all types of samples within micro-tubes. Whether gently mixing, extracting RNA/DNA or destroying the cell wall, these packages can provide fast and complete effective results. Sample volumes as small as 0.03mL can be easily processed in a matter of seconds and cleanup is just as easy. Please contact us for 220 volt homogenizing packages.CLS-5034-16 Premium Micro Package Contains:1 each CLS-5030-10 Homogenizer1 each CLS-5032-10 5 x 75mm Generator1 each CLS-5032-11 7 x 75mm Generator1 each CLS-5030-30 Stand Assembly