Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Premium Microscope Slide Box with Cork Lining

Catalog No. 034497 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Red
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-449-7 Red
03-449-8 Blue
03-348-10 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-449-7
Only null left
Add to Cart
Add to Cart

Store and transport slides safely with durable ABS cases. Fisherbrand™ Colored ABS Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage.

  • Molded, rigid boxes securely hold slides in grooved slots
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Friction-fit cover closes easily and allows for easy access to slides
  • Features a thumb-latch lock mechanism for additional security
  • Stackable

Specifications

Type Slide Box
Slide Capacity 25
Closure Type Friction-Fit Lock
Color Red
Device Holds 25 Slides
Dimensions (L x W x H) 141 x 88 x 35 mm
For Use With (Application) Safely storing or transporting microscope slides
For Use With (Equipment) Standard 25 x 75 mm (1 x 3 in.) Microscope Slides
Includes Slide Inventory Sheet
Material ABS
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Lining Material Cork
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.