Learn More
Glo Germ Company Premium Mini Kit
SDP
Catalog No. NC9023120
Encompass_Preferred
Premium Mini Kit. Introducing The Premium Mini Kit Which Includes The 1003 Lamp W/Battery, A 2 Oz. Bottle Of Glo Germ Gel, A 1.2 Oz Glo Germ Powder, Trainers Manual And A Set Of Activity Sheets. This Kit Is Excellent For Handwashing Training (Up To 50 Demonstrations) And Cross Contamination Demonstration And Testing (Over 50 Demonstrations).
Catalog No. NC9023120 Supplier Glo Germ Company Supplier No. PMK
Premium Mini Kit. Introducing The Premium Mini Kit Which Includes The 1003 Lamp W/Battery, A 2 Oz. Bottle Of Glo Germ Gel, A 1.2 Oz Glo Germ Powder, Trainers Manual And A Set Of Activity Sheets. This Kit Is Excellent For Handwashing Training (Up To 50 Demonstrations) And Cross Contamination Demonstration And Testing (Over 50 Demonstrations).