Learn More
Surgical Design™ Premium Pederson Vaginal Speculum
Catalog No. 21125071 Shop All Surgical Design Products
Change view
Click to view available options
Size:
Large
Medium
Narrow
Small
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-125-071 Large
21-125-068 Small
21-125-069 Medium
21-125-070 Narrow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-125-071 Supplier Surgical Design™ Supplier No. ORG278
Add to Cart
Add to Cart
Pederson vaginal speculum features a slim profile for comfortable, controlled visualization during gynecological examinations.
  • Available Sizes: Small, Medium, Large, Narrow
  • Narrow, tapered blades ideal for patients with smaller introitus or vaginal stenosis
  • Smooth screw-lock mechanism for precise and stable opening
  • Rounded, polished edges enhance patient comfort during exams
  • Crafted from medical-grade stainless steel—durable, corrosion-resistant, and fully autoclavable
  • Suitable for OB/GYN, primary care, emergency medicine, and outpatient procedures
Specifications
Product Type Vaginal Speculum
Grade Medical
Size Large
Autoclavable Autoclavable