Learn More
Medicus Health Premium Phlebotomy Cart with Adjustable Height
SDP
Catalog No. NC3595097
Encompass
• The Medicus "Best in Class" Phlebotomy Cart has many unique features that have been developed from years of customer feedback including integrated cast aluminum polished push handle, easy touch height adjustment and compact base footprint for easy storage
• Phlebotomy Cart is lightweight but durable with premium casters making it very easy to push from room to room
• Fingertip control adjusts cart surface 18" in height from 29" to 47" from the floor to be at the height you need for working or for pushing the cart
• Unique counter weight base with platinum finish prevents cart from tipping and gives it a smooth ride. Base measures: 13.25”W x 14”D x .25”H
• Height Adjustment Hand Lever
• Integrated contoured handle is ergonomically shaped to aid in the movement of the cart
• Includes Utility Basket to hold supplies that measures: 12” W x 5”D x 4”H.
• Includes standard 8 Quart Sharps Container on wire frame
Catalog No. NC3595097 Supplier Medicus Health Supplier No. 5271M2
• The Medicus "Best in Class" Phlebotomy Cart has many unique features that have been developed from years of customer feedback including integrated cast aluminum polished push handle, easy touch height adjustment and compact base footprint for easy storage
• Phlebotomy Cart is lightweight but durable with premium casters making it very easy to push from room to room
• Fingertip control adjusts cart surface 18" in height from 29" to 47" from the floor to be at the height you need for working or for pushing the cart
• Unique counter weight base with platinum finish prevents cart from tipping and gives it a smooth ride. Base measures: 13.25”W x 14”D x .25”H
• Height Adjustment Hand Lever
• Integrated contoured handle is ergonomically shaped to aid in the movement of the cart
• Includes Utility Basket to hold supplies that measures: 12” W x 5”D x 4”H.
• Includes standard 8 Quart Sharps Container on wire frame