Learn More
Gilson™ Premium Pipette Service for Single Channel Pipettes
Catalog No. p-7184957 Shop All Gilson Products
Change view
Click to view available options
Product Type:
ISO Pipette Premium Lifetime Service
Pipette Premium Lifetime Service
Service Offering:
After Repair Calibration
As-found/After Repair Calibration
Calibration Points:
2 x 2 Measurements
4 x 2 Measurements
5 x 3 Measurements
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Service Offering Calibration Points
SVC101235G ISO Pipette Premium Lifetime Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101259G ISO Pipette Premium Lifetime Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101223G Pipette Premium Lifetime Service As-found/After Repair Calibration 4 x 2 Measurements
SVC101247G Pipette Premium Lifetime Service As-found/After Repair Calibration 5 x 3 Measurements
SVC101199G Pipette Premium Lifetime Service After Repair Calibration 2 x 2 Measurements
SVC101211G Pipette Premium Lifetime Service After Repair Calibration 4 x 2 Measurements
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SVC101235G Supplier Gilson™ Supplier No. SVC101235
Only null left
Add to Cart
Add to Cart
Gilson Premium Pipette Service is the most comprehensive level of service, providing users with reliable performance and predictable pricing. At least once per year Gilson comes with a lifetime warranty on the pipette and all necessary repairs are included at no additional cost.
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • Calibration and Detailed Certificate
  • ISO 17025 Calibration Available
  • Lifetime Warranty
  • Free Repairs
  • 3 day turn around time
Specifications
For Use With (Equipment) Single-channel Pipette
Certifications/Compliance ISO 17025 Accredited Service
Description Data collected before and after repair
Calibration Points 4 x 2 Measurements
Duration 3 Days
Product Type ISO Pipette Premium Lifetime Service
Service Offering As-found/After Repair Calibration