Learn More
Mopec™ Premium Platform Scale
SDP
Catalog No. 22445363 Shop All Mopec Products
Change view
Click to view available options
Capacity:
2200 g
Catalog No. Capacity
22-445-363 2200 g
1 options
Catalog No. 22-445-363 Supplier Mopec™ Supplier No. BB165
Add to Cart
Add to Cart
Ideal for laboratory and industrial applications, the Platform Scale features an easy to navigate color touchscreen display and slim design, allowing for stackable storage. Geared for high performance with fast stabilization time and high resolution weighing results.
  • Includes overload protection and expanded connectivity options
  • RS232 Interface
  • Allows data connection to PC or printer
  • AC adapter included
  • GLP/GMP Data capabilities including date, time, balance serial number, user ID/project ID, and results
Specifications
Capacity 2200 g
Display Type Digital