Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Premium Plus Replacement Inventory Cards
GREENER_CHOICE

Catalog No. 14127569 Shop All Fisherbrand Products
Change view
Click to view available options
Type: Inventory Card
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14-127-569 Type: Inventory Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-569
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Premium Plus Replacement Inventory Cards designed to fit into the premium plus slide boxes.

  • Replacement inventory cards for use with Premium Plus Microscope Slide Boxes

Specifications

Type Inventory Card
Slide Capacity 100
Autoclavable Not Autoclavable
Color Gray
Compartments 100
For Use With (Application) Histology
Height (English) 11 in.
Height (Metric) 28 cm
Length (English) 8.5 in.
Length (Metric) 21.6 cm
Material Cardboard
Quantity 10 Pk.
Shape Rectangular
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.