Learn More
Bellco Glass, Inc. Premium Pouring Ring, 45mm Clear
SDP
Catalog No. 501945758
Encompass_Preferred
Provides drip free pouring and protects bottle rim from chipping. MADE IN THE USA Autocavable Size: 45mm Color: clear.
Catalog No. 50-194-5758 Supplier Bellco Glass, Inc. Supplier No. 563700041
Provides drip free pouring and protects bottle rim from chipping. MADE IN THE USA Autocavable Size: 45mm Color: clear.