Learn More
Med Vet International Premium Small Animal Dental Scaler/Probe Kit Handles - Kit Stainless Handle, J1513, Each
Catalog No. 5000585785
Encompass_Preferred
Kit includes: • Periosteal Elevator Dual end 2mm and 4 mm • Resorptive Lesion Probe • Sharp Spoon Small Scaler • Sickle Scaler Normal • Exploring and Measuring Probe Dual End • Jacquette Supra Gingival Scaler • McCall Scaler Dual End • Scaler On & Off Side • Incisor Luxators Double End • Explorer Scaler Double End • Spatula Double End • Deciduous Elevator Small (2mm) • Deciduous Elevator Medium (3mm) • Examination Single End Mirror • Periosteal Elevator Curved Ended
Catalog No. 50-005-85785 Supplier Med Vet International Supplier No. JOR1513
Kit includes: • Periosteal Elevator Dual end 2mm and 4 mm • Resorptive Lesion Probe • Sharp Spoon Small Scaler • Sickle Scaler Normal • Exploring and Measuring Probe Dual End • Jacquette Supra Gingival Scaler • McCall Scaler Dual End • Scaler On & Off Side • Incisor Luxators Double End • Explorer Scaler Double End • Spatula Double End • Deciduous Elevator Small (2mm) • Deciduous Elevator Medium (3mm) • Examination Single End Mirror • Periosteal Elevator Curved Ended