Learn More
Four E's Healthcare Premium SMS Lab Coat with Anti-Static Coating
Catalog No. 22021306 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-021-306 2X-Large
22-021-302 Small
22-021-303 Medium
22-021-304 Large
22-021-305 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-021-306
Premium knee-length lab coat from Four E's Healthcare made of 3-layer breathable SMS fabric with anti-static coating, offers superior comfort for extended use and protects from low fluid levels
  • Anti-static SMS lab coats with knit collars, cuffs and snap front with five buttons
  • Protective 3-ply breathable SMS fabric for comfort and protection
  • Offers protection from low fluid levels
    • SMS fabric material meets AAMI Level 2 liquid barrier performance
  • Color: blue
  • Three reinforced pockets (two hip pockets, one breast pocket)
  • Comes with anti-static coating
    • Keeps wearer safe in static-prone environments and avoids clinging
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Fluids
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Model 4ESMSLC45
Certifications/Compliance AAMI Level 2
Construction 3-ply SMS fabric with anti-static coating
Show More Show Less
Please note: The SMS fabric is fluid resistant, and meets AAMI Level 2 Liquid barrier performance. However, due to the snap front design, the coat does not qualify for AAMI PB70 standard testing.