Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Premium Split Cowhide Leather Palm Gloves with Aramid Fiber Stitching

Catalog No. 1146260C Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-462-60C X-Large
11-462-60B Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-462-60C Supplier Wells Lamont™ Supplier No. Y3015XL
  • $347.00
    $196.65 / Dozen of 12

    Save $150.35

    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,203.00
    $683.65 / Case of 4 DZ

    Save $519.35

    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premium Split Cowhide Leather Palm Gloves with Aramid Fiber Stitching are constructed from superior quality grade leather, and stitched with aramid fibers for increased strength and durability.

  • Sewn with aramid fiber thread for increased strength at the seams
  • Premium quality split cowhide extends the glove's usage cycle
  • An extended gauntlet cuff provides protection up to the forearm
  • Leather knuckle strap
  • Canvas back material
  • Wing thumb

Specifications

Back Material Canvas
Construction Cut and Sewn
Cuff Material Canvas
Color Natural, Gray
Grade Superior
Hand Specific
Material Shoulder Split Cowhide Leather, Aramid Fiber
Product Type Leather Palm Gloves
Size X-Large
Style Gunn
For Use With (Application) Steel Processing, Metal Handling, Construction
Gender Unisex
Glove Cuff Color Natural
Glove Cuff Style Gauntlet
Grip Design Smooth
Hem Color Blue
Lining Unlined
Palm Type Leather
Series Y3015
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.