Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen™ Clear Contrast Clarifier Differentiator

Catalog No. 23730612 Shop All Scigen Inc Products
Change view
Click to view available options
Quantity:
1/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-730-612 1/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-730-612 Supplier Scigen™ Supplier No. 7119
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A water based ready to use differentiator which can be used in manual staining or on auto-strainers.

Scigen™ Premium Clear Contrast Differentiator is designed to to produce cell transparency and remove any nonspecific hematoxylin staining. Our Clear Contrast Differentiator is water based ready to use differentiator with can be used in manual staining or on auto-strainers. Most differentiators or clarifying agents are alcohol based; evaporation of the alcohol causes inconsistent nuclear staining results.

After hematoxylin staining, excess hematoxylin is rinsed off the tissue sections, slides are differentiated in Scigen™ Premium Clear Contrast Differentiator for 15 seconds to 1 minute depending on hematoxylin staining time and background staining. Scigen™ Premium Clear Contrast Differentiator is not a concentrate and requires no dilution.

Specifications

Color Clear, Water white
Description Provides clear, precise differentiation and contrast between hematoxylin and eosin
For Use With (Application) Solution for the differentiation of hematoxylin stains
Includes Staining Protocol
Material Aqueous Based
Progressive/Regressive Regressive
Quantity 1/Cs.
Solution Type Aqueous
Storage Requirements Room Temperature
Synonym Differentiating
Product Type Differentiating Solution
Show More Show Less

FOR IN VITRO LABORATORY USE POISON: NOT FOR DRUG, FOOD, OR HOUSEHOLD USE.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.