Learn More
Yamazen Science Inc Disposable Silica Gel Packed Column, with High Throughput Purification, Luer Lock Type
Catalog No. NC1465391
Encompass
This item has a minimum qty of 2 per supplier requirements.
Premium Universal Column: High Throughput Purification PerformancePore Size 60 angstroms, Particle Size 25/40um, Silica Gel, Luer Lock TypeSize: L10/pk
Catalog No. NC1465391 Supplier Yamazen Science Inc Supplier No. UW10301
This item has a minimum qty of 2 per supplier requirements.
Premium Universal Column: High Throughput Purification PerformancePore Size 60 angstroms, Particle Size 25/40um, Silica Gel, Luer Lock TypeSize: L10/pk