Learn More
Yamazen Science Inc Premium Universal Column, Silica Gel, Luer Lock Type, 10/Pack
Catalog No. NC1465384
Encompass
This item has a minimum qty of 3 per supplier requirements.
Premium Universal Column: High Throughput Purification PerformancePore Size 60 angstroms, Particle Size 25/40um, Silica Gel, Luer Lock TypeSize: S10/pk
Catalog No. NC1465384 Supplier Yamazen Science Inc Supplier No. UW10901
This item has a minimum qty of 3 per supplier requirements.
Premium Universal Column: High Throughput Purification PerformancePore Size 60 angstroms, Particle Size 25/40um, Silica Gel, Luer Lock TypeSize: S10/pk