Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC VP Racing Fuels Engine Fuel, 50:1, 94 Octane, 54 gal

Catalog No. 502868993
Encompass

VP Racing Fuels premixed small engine fuel is designed for outdoor power equipment. This fuel is formulated for 2-cycle engines, providing a 50:1 premix for optimal performance and protection.

  • Premixed 50:1 fuel type
  • Compatible with 2 cycle engines
  • Contains no ethanol
  • Octane rating: 94
  • Container size: 54 gal
  • Flash point: -31.9 F
  • Specific gravity: 0.7225

Catalog No. 50-286-8993 Supplier MSC Supplier No. 28990778
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VP Racing Fuels premixed small engine fuel is designed for outdoor power equipment. This fuel is formulated for 2-cycle engines, providing a 50:1 premix for optimal performance and protection.

  • Premixed 50:1 fuel type
  • Compatible with 2 cycle engines
  • Contains no ethanol
  • Octane rating: 94
  • Container size: 54 gal
  • Flash point: -31.9 F
  • Specific gravity: 0.7225

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.