Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC VP Racing Fuels 2-Cycle, 40:1 Premixed, 94 Octane, 54 gal

Catalog No. 502816746
Encompass

VP Racing Fuels Outdoor Power Equipment Fuel is a specially formulated, ethanol-free fuel designed for 2-cycle small engines, ensuring optimal performance and protection.

  • Premixed 40:1 ratio for convenience
  • Ethanol-free formulation prevents fuel system damage
  • Provides 94 octane for enhanced engine performance
  • Suitable for 2-cycle engines
  • Flash point of -31.9 °F
  • Specific gravity of 0.7225

Catalog No. 50-281-6746 Supplier MSC Supplier No. 28990901
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VP Racing Fuels Outdoor Power Equipment Fuel is a specially formulated, ethanol-free fuel designed for 2-cycle small engines, ensuring optimal performance and protection.

  • Premixed 40:1 ratio for convenience
  • Ethanol-free formulation prevents fuel system damage
  • Provides 94 octane for enhanced engine performance
  • Suitable for 2-cycle engines
  • Flash point of -31.9 °F
  • Specific gravity of 0.7225

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.