Learn More
Takara Bio Premix Ex Taq™ DNA Polymerase (Perfect Real Time), 200 Rxns
SDP
Catalog No. 50443999
Encompass
Premix Ex Taq (Perfect Real Time) is designed for probe-based qPCR, and is also suitable for fast PCR. Premix Ex Taq allows accurate target quantification and detection over a broad dynamic range and makes it possible to obtain highly reproducible and reliable realtime PCR results. The product is supplied as a 2X premix to facilitate easy preparation of reaction mixtures. The combination of Takara Ex Taq HS, a hot start PCR enzyme that uses an anti-Taq antibody, and a buffer optimized for real-time PCR suppresses non-specific amplification and allows high amplification efficiency and high detection sensitivity in real-time PCR analyses. 200 Rxns
Catalog No. 50-443-999 Supplier Takara Bio Supplier No. RR039A
Premix Ex Taq (Perfect Real Time) is designed for probe-based qPCR, and is also suitable for fast PCR. Premix Ex Taq allows accurate target quantification and detection over a broad dynamic range and makes it possible to obtain highly reproducible and reliable realtime PCR results. The product is supplied as a 2X premix to facilitate easy preparation of reaction mixtures. The combination of Takara Ex Taq HS, a hot start PCR enzyme that uses an anti-Taq antibody, and a buffer optimized for real-time PCR suppresses non-specific amplification and allows high amplification efficiency and high detection sensitivity in real-time PCR analyses. 200 Rxns