Learn More
Takara Bio Premix Ex Taq™ DNA Polymerase Hot Start Version 100 Rxns
SDP
Catalog No. 50443998
Encompass
A hot-start, 2X PCR master mix containing Takara Ex Taq DNA polymerase, optimized buffer, and dNTPs. Please refer to the User Manual for buffer composition. Takara Ex Taq DNA polymerase combines the proven performance of Takara Taq polymerase with the proofreading activity of an efficient 3'-to-5' exonuclease, for high-sensitivity, high-efficiency PCR. Ex Taq is optimized for amplicons up to 20 kb from genomic DNA, and up to 30 kb from lambda DNA. 100 Rxns
Catalog No. 50-443-998 Supplier Takara Bio Supplier No. RR030A
A hot-start, 2X PCR master mix containing Takara Ex Taq DNA polymerase, optimized buffer, and dNTPs. Please refer to the User Manual for buffer composition. Takara Ex Taq DNA polymerase combines the proven performance of Takara Taq polymerase with the proofreading activity of an efficient 3'-to-5' exonuclease, for high-sensitivity, high-efficiency PCR. Ex Taq is optimized for amplicons up to 20 kb from genomic DNA, and up to 30 kb from lambda DNA. 100 Rxns