Learn More
World Precision Instrument Premixslip Instrument Lubricant and Rust Inhibitor, 16 oz.
SDP
Catalog No. NC1510635
Encompass
A 10-minute soak keeps surgical instruments looking new
Premixslip is a non-sticky, non-toxic, silicone-free and water soluble mineral oil lubricant that can withstand the high temperatures of autoclaving without becoming gummy. Forming a protective barrier on surgical instruments, Premixslip prevents rusting, staining and spotting while restoring articulation to sticky box-locks and joints.
Catalog No. NC1510635 Supplier World Precision Instrument Supplier No. WP34552
A 10-minute soak keeps surgical instruments looking new
Premixslip is a non-sticky, non-toxic, silicone-free and water soluble mineral oil lubricant that can withstand the high temperatures of autoclaving without becoming gummy. Forming a protective barrier on surgical instruments, Premixslip prevents rusting, staining and spotting while restoring articulation to sticky box-locks and joints.