Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Premo Flex, 3/4"" x 1.15""; 50 ft/pk
Catalog No. NC1694865
Encompass
Masterflex Premp Flex transfer tubing is ideal for use with fuels such as petroleum oils, gasoline, kerosene, and lubricating oils, while maintaining tube integrity. The Buna-N tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube meets RMA Class A high oil resistance and nonconductive at 1000 VDC certification. The tube cover meets Class B medium oil resistance and nonconductive at 1000 VDC certification.
Catalog No. NC1694865 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9595305
Masterflex Premp Flex transfer tubing is ideal for use with fuels such as petroleum oils, gasoline, kerosene, and lubricating oils, while maintaining tube integrity. The Buna-N tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube meets RMA Class A high oil resistance and nonconductive at 1000 VDC certification. The tube cover meets Class B medium oil resistance and nonconductive at 1000 VDC certification.