Learn More
Andwin Scientific Wipers Premoistened with Isopropyl Alcohol/Deionized Water, 100/pk
SDP
Catalog No. NC0661008
Encompass_Preferred
Premoistened wipers are designed for environmentally sensitive areas, and are ideal for wiping optical parts, delicate glassware, electrodes, cuvettes, microscopes, and lenses.
This item is not returnable. View return policy
Catalog No. NC0661008 Supplier Andwin Scientific Supplier No. 21910110
Premoistened wipers are designed for environmentally sensitive areas, and are ideal for wiping optical parts, delicate glassware, electrodes, cuvettes, microscopes, and lenses.