Learn More
Leica Biosystems PREMR COVER GLASS ACS 24X40X1
Catalog No. NC1532260
Encompass_Preferred
Premier Cover Glass has been specially designed to work with automated coverslippers. Extra processes to prevent sticking are employed during manufacturing. Premier coverglass available in multiple sizes. Thinness: 013mm - 0.17mm Sizes range from 22 X 40mm to 24 x 60mm as noted below.
Catalog No. NC1532260 Supplier Leica Biosystems Supplier No. 3800140ACS
Premier Cover Glass has been specially designed to work with automated coverslippers. Extra processes to prevent sticking are employed during manufacturing. Premier coverglass available in multiple sizes. Thinness: 013mm - 0.17mm Sizes range from 22 X 40mm to 24 x 60mm as noted below.