Learn More
Perkin Elmer US LLC Pharmed Three-Stop Peristaltic Pump Tubing - 1.14 mm I.D., Red/Red/Red, Pkg. 6
Catalog No. 509052083
Encompass_Preferred
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. It can also be used with the AS9X Autosampler and ships as a package of 6.
Catalog No. 50-905-2083 Supplier Perkin Elmer US LLC Supplier No. B3140730
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. It can also be used with the AS9X Autosampler and ships as a package of 6.