Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prenisteine | 5287-46-7 | 99.8% | 250 MG
SDP

Catalog No. 5000232420
Encompass_Preferred

Prenisteine is an expectorant agent supplied as a high-purity research reagent for laboratory studies and analytical reference.

  • Purity: 99.8%.
  • Chemical formula: C8H15NO2S.
  • Molecular weight: 189.28 g/mol.
  • Available in multiple pack sizes including 250 MG.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-32420 Supplier Medchemexpress LLC Supplier No. HY121553250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prenisteine is an expectorant agent supplied as a high-purity research reagent for laboratory studies and analytical reference.

  • Purity: 99.8%.
  • Chemical formula: C8H15NO2S.
  • Molecular weight: 189.28 g/mol.
  • Available in multiple pack sizes including 250 MG.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.