Learn More
Cayman Chemical Prenylamine, Purity: ≥98%, CAS: 390-64-7, 100 mg
SDP
Catalog No. 502989365
Encompass_Preferred
A calcium channel inhibitor; inhibits PMCA in isolated and purified pig cardiac sarcolemma; binds to a hydrophobic site on calcium-bound calmodulin (CaM) with a Kd value of 0.5 µM and inhibits CaM-activated cAMP PDE activity at 10-50 µM, an effect that is negatively associated with the concentration of calmodulin; shortens action potentials and decreases the inward calcium current in single guinea pig ventricular myocytes in the absence and presence of propranolol and phentolamine at 30 μM; decreases epinephrine, serotonin, and dopamine levels in rat brain at 50 mg/kg; decreases epinephrine levels in rat heart; protects anesthetized rats from coronary artery occlusion-induced arrhythmia at 0.5 mg/kg but increases mortality due to atrioventricular block leading to asystole at 5 mg/kg
Catalog No. 50-298-9365 Supplier Cayman Chemical Supplier No. 29430100 MG
A calcium channel inhibitor; inhibits PMCA in isolated and purified pig cardiac sarcolemma; binds to a hydrophobic site on calcium-bound calmodulin (CaM) with a Kd value of 0.5 µM and inhibits CaM-activated cAMP PDE activity at 10-50 µM, an effect that is negatively associated with the concentration of calmodulin; shortens action potentials and decreases the inward calcium current in single guinea pig ventricular myocytes in the absence and presence of propranolol and phentolamine at 30 μM; decreases epinephrine, serotonin, and dopamine levels in rat brain at 50 mg/kg; decreases epinephrine levels in rat heart; protects anesthetized rats from coronary artery occlusion-induced arrhythmia at 0.5 mg/kg but increases mortality due to atrioventricular block leading to asystole at 5 mg/kg