Learn More
Microsolv Technology Corporation Prep HPLC Column, HPS Phenyl, 5um, 21.2mm ID x 50mm Length, 120A. 1 EA.
Supplier:  Microsolv Technology Corporation 75020P50
Encompass_Preferred
This Cogent HPS Phenyl™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is fully end capped and is manufactured with a mono functional, 4 carbon ligand that has an attached Phenyl group and it is bonded to the highest purity, 120A silica available. With a 5µm Particle, this Column works well with higher flow rates producing lower back pressures compared to columns with smaller particles making ideal for the purification of Isomers (positional), Tocopherols, Flavonoids, Polynuclear Aromatics, Nitro-Aromatic Compounds, Active Pharmaceutical Ingredients (APIs), Natural Products and Related Compounds in Reversed Phase HPLC. A specialized HPLC System is required for columns this size. Compare this column's performance to Luna ™ or Inertsil ™ columns.
Catalog No. 50-227-1044