Learn More
Microsolv Technology Corporation Prep HPLC Column, Phenyl Hydride, 4um, 100A. 21.2mm ID x 150mm Length, Preparative. 1 EA.
Supplier:  Microsolv Technology Corporation 69020P21150
Encompass_Preferred
This Cogent Phenyl Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for purification of compounds with aromatic rings that are difficult to retain or separate in Reversed Phase, including aromatic amines, small peptides and azides. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0876
Add to Cart
Request a Quote