Learn More
Microsolv Technology Corporation Prep HPLC Column, RP C18, 10um, 100A. 30mm ID x 50mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68018P3050
Encompass_Preferred
This Cogent RP C18™ Stationary Phase has been a premium value, and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. The 10 µm Particle works well with higher flow rates and produces lower back pressures than smaller particles. The extra large diameter of this column provides very high loading capacity which is great for purification of hydrophobic or neutral compounds making this it ideal for Preparative HPLC. A specialized HPLC System is required for columns this size.
Catalog No. 50-226-9818