Learn More
Microsolv Technology Corporation Prep HPLC Column, RP C18, 5um, 100A. 21.2mm ID x 250mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68518P21250
Encompass_Preferred
This Cogent RP C18™ Stationary Phase has been a premium value, and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. The 5 µm Particle works well with prep flow rates and produces lower back pressures than smaller particles. The large diameter and extra length of this column provides high loading capacity which is great for purification of hydrophobic or neutral compounds making this it ideal for Preparative HPLC. A specialized HPLC System is required for columns this size.
Catalog No. 50-226-9867
Request a Quote
Please call customer service for current pricing. 1-800-766-7000