Learn More
Microsolv Technology Corporation Prep HPLC Column, UDC Cholesterol, 4um, 100A. 30mm ID x 50mm Length, Preparative. 1 EA.
Supplier:  Microsolv Technology Corporation 69069P3050
Encompass_Preferred
This UDC Cholesterol™ HPLC is made with Cogent TYPE-C™ Silica , and provides unique Selectivity due to different surface chemistry, compared to typical Reversed Phase Columns. This Silica Hydride Phase is modified with an 11carbon chain with a cholesterol molecule attached at the terminus. This gives this column the ability to form “slots” and separate some compounds based on shape. Also, due to the direct silicon carbon bonds, this column has no need for end-capping. Use in Reversed Phase, Normal Phase and Aqueous Normal Phase (ANP). Chosen for analyzing or collecting steroidal and other hard to retain compounds including some lipids, peptides, proteins & oligonucleotides. Faster Equilibration than other Reversed Phase or HILIC Columns Very Hydrophobic Robust Column Lifetimes Temperature and Solvent Changes Selectivity A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0908
Add to Cart
Request a Quote