Learn More
CROSSTEX INTERNATIONAL Prepared Growth Media For Use With Bacillus atrophaeus
Catalog No. NC1690631
Encompass_Preferred
The prepared growth media consists of an exclusively formulated Tryptic Soy Broth (TSB) modified with pH indicator. The pH indicator causes a transition in media color from green to yellow when Bacillus atrophaeus bacterial growth is present, allowing for a reduced incubation when used in conjunction with Crosstex’s Biological Indicators (BIs) and Inoculated Carriers for EO and Dry Heat sterilization processes.
This item is not returnable. View return policy
Catalog No. NC1690631 Supplier CROSSTEX INTERNATIONAL Supplier No. NMGMBTB100
The prepared growth media consists of an exclusively formulated Tryptic Soy Broth (TSB) modified with pH indicator. The pH indicator causes a transition in media color from green to yellow when Bacillus atrophaeus bacterial growth is present, allowing for a reduced incubation when used in conjunction with Crosstex’s Biological Indicators (BIs) and Inoculated Carriers for EO and Dry Heat sterilization processes.