Learn More
G-Biosciences Prep Desalting Column

Description
- Prep Desalting columns are mostly used for removal of salt or buffer exchange before or after different chromatographic steps.
- Prep Desalting columns can be used for separation of proteins based on their sizes.
- G‐Biosciences offer different sizes of desalting columns.
- Prep Desalting columns are made of biocompatible polypropylene, which does not interact with biomolecules.
- The column has a stopper at the inlet and one top plug, one end cap, two filter discs.
Specifications
Specifications
| Product Type | Desalting Column |
| Format | Gravity Flow Column |
| Column Type | Size-exclusion, Proprietary Resin |
| MWCO | 5.0 kDa |
| Product Line | Pierce |
| Disposable | Reusable |
| Quantity | 1 x 150 mL |
| Sample Volume (Metric) | 150 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.