Learn More
Waters Corp Waters Prep Funnel Collection Rack, 128 Bottle Capacity, Fraction Collector III Compatible
Catalog No. 50793112 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Prep Funnel Collection Rack is an optional accessory designed for use with the Waters Fraction Collector III. It replaces the standard tube rack to facilitate the collection of larger volume fractions.
  • Collects larger volume fractions.
  • Accommodates up to four prep funnels.
  • Each funnel supports 32 fractions.
  • Enables collection in up to 128 large bottles or flasks.
  • Compatible with an optional tabletop stand.
Catalog No. 50-793-112 Supplier Waters Corp Supplier No. 725000148
The Waters Prep Funnel Collection Rack is an optional accessory designed for use with the Waters Fraction Collector III. It replaces the standard tube rack to facilitate the collection of larger volume fractions.
  • Collects larger volume fractions.
  • Accommodates up to four prep funnels.
  • Each funnel supports 32 fractions.
  • Enables collection in up to 128 large bottles or flasks.
  • Compatible with an optional tabletop stand.