Learn More
Microsolv Technology Corporation Prep Guard Column Cartridge, RP C18, 5um, 100A. 30mm ID x 10mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 68518P30PG1
Encompass_Preferred
This Guard Column Cartridge is packed with 5µm, Cogent RP C18™ Stationary Phase. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. It is designed to protect your RP C18 Prep Column from particulate and contaminants that will adsorb to it. Packed in a Stainless Steel Tube, it has SS Frits. Use this Guard Column Cartridge with a 30mm ID Prep Column Holder.
Catalog No. 50-227-0203