Learn More
Med Vet International Skin Prep Solution 3M Duraprep 6 mL Foam Applicator 0.7% / 74% Strength Iodine Povacrylex / Isopropyl Alcohol Nonsterile, 8635, Each
Catalog No. 502832378
Encompass_Preferred
3M™ Duraprep™ surgical solution comes in a convenient 6 mL applicator for quick, efficient skin coverage It forms a thick layer of film that quickly kills bacteria and continues to prevent microbial build-up When dry, it stays in place even during operations and other procedures This film serves as a primer that holds incise drapes securely in place Duraprep™ 6 mL reduces the chance of infection The film is easy to remove using saline or a sponge
Catalog No. 50-283-2378 Supplier Med Vet International Supplier No. 202719EA
3M™ Duraprep™ surgical solution comes in a convenient 6 mL applicator for quick, efficient skin coverage It forms a thick layer of film that quickly kills bacteria and continues to prevent microbial build-up When dry, it stays in place even during operations and other procedures This film serves as a primer that holds incise drapes securely in place Duraprep™ 6 mL reduces the chance of infection The film is easy to remove using saline or a sponge