Learn More
Analytical Sales & Services Prep Mobile Phase Safety Cap with 1 Port for 1/8" OD Tubing, 1 Port for 1/4" OD Tubing
Catalog No. 5000805335
Encompass
Prep Mobile Phase Safety Cap with One Port for 1/8" OD Tubing, One Port for 1/4" OD Tubing, and One Prep Air Check Valve\n\nSuggested for Flow Rates >100mL and <500mL\n\nCap Thread Size: S55
Catalog No. 50-008-05335 Supplier Analytical Sales & Services Supplier No. CCP1418
Prep Mobile Phase Safety Cap with One Port for 1/8" OD Tubing, One Port for 1/4" OD Tubing, and One Prep Air Check Valve\n\nSuggested for Flow Rates >100mL and <500mL\n\nCap Thread Size: S55