Learn More
Waters Corp WATERS CORPORATION Nova-Pak C18 Radial-Pak, 6 μm, 40 mm x 100 mm
Catalog No. 50785436 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 μm particle technology. They offer efficient and fast chromatography with higher resolution.
  • Offers higher resolution compared to a 5 μm column.
  • More efficient separations compared to a 5 μm column.
  • Faster chromatography compared to a 5 μm column.
Catalog No. 50-785-436 Supplier Waters Corp Supplier No. WAT037704
Add to Cart
Add to Cart
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 μm particle technology. They offer efficient and fast chromatography with higher resolution.
  • Offers higher resolution compared to a 5 μm column.
  • More efficient separations compared to a 5 μm column.
  • Faster chromatography compared to a 5 μm column.