Learn More
Waters Corp Waters Nova-Pak Prep Column, C18, 6 μm, 7.8 x 300 mm
Catalog No. 50785210 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 μm particle technology. These columns offer higher resolution, more efficient separations, and faster chromatography compared to a 5 μm column.
  • Silica-based, semi-preparative, and preparative columns
  • Based on 6 μm particle technology
  • Offers higher resolution, more efficient separations, and faster chromatography
Catalog No. 50-785-210 Supplier Waters Corp Supplier No. WAT025820
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 μm particle technology. These columns offer higher resolution, more efficient separations, and faster chromatography compared to a 5 μm column.
  • Silica-based, semi-preparative, and preparative columns
  • Based on 6 μm particle technology
  • Offers higher resolution, more efficient separations, and faster chromatography