Learn More
Waters Corp Nova-Pak C18 Radial-Pak Guard, 60Å, 6 µm, 40 mm X 10 mm, 2/pk
Catalog No. 50785446 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 µm particle technology. Compared to a 5 µm column, Nova-Pak HR columns offer higher resolution, more efficient separations, and faster chromatography.
Catalog No. 50-785-446 Supplier Waters Corp Supplier No. WAT037854
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 µm particle technology. Compared to a 5 µm column, Nova-Pak HR columns offer higher resolution, more efficient separations, and faster chromatography.