Learn More
Waters Corp Waters Prep Column, Silica, 6 μm, 7.8 x 300 mm
Catalog No. 50785211 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Nova-Pak Silica Prep Column is a semi-preparative and preparative column designed for normal-phase HPLC applications. Featuring 6 μm silica particles, a 60 Å pore size, and dimensions of 7.8 mm x 300 mm, it offers higher resolution, more efficient separations, and faster chromatography compared to 5 μm columns.
  • Normal-phase separation mode
  • Spherical particle shape
  • Silica bonding technology
  • High silanol activity
  • pH range of 2 to 8
  • Temperature limits up to 45 °C
  • Maximum pressure of 6000 psi (415 bar)
  • Not endcapped
  • Suitable for HPLC Preparative systems and LC/MS, LC techniques
  • Offers higher resolution, more efficient separations, and faster chromatography
Catalog No. 50-785-211 Supplier Waters Corp Supplier No. WAT025821
The Nova-Pak Silica Prep Column is a semi-preparative and preparative column designed for normal-phase HPLC applications. Featuring 6 μm silica particles, a 60 Å pore size, and dimensions of 7.8 mm x 300 mm, it offers higher resolution, more efficient separations, and faster chromatography compared to 5 μm columns.
  • Normal-phase separation mode
  • Spherical particle shape
  • Silica bonding technology
  • High silanol activity
  • pH range of 2 to 8
  • Temperature limits up to 45 °C
  • Maximum pressure of 6000 psi (415 bar)
  • Not endcapped
  • Suitable for HPLC Preparative systems and LC/MS, LC techniques
  • Offers higher resolution, more efficient separations, and faster chromatography