Learn More
Waters Corp Porasil Silica Radial-Pak Column, 125Å, 55 - 105 µm, 8 mm X 100 mm, 1/pk
Supplier:  Waters Corp WAT025851
Encompass_Preferred
Waters Porasil 15 - 20µm silica packing material provides long column life and low backpressure. The packing also has a high surface area making it ideal for preparative work.
Catalog No. 50-785-227