Learn More
Zymo Research Corporation Prep Solution (30 ml)
SDP
Catalog No. 501251610
Encompass_Preferred
This prepares the columns or plates for the efficient removal of PCR inhibitors such as polyphenolics, humic/fulvic acids, tannins, melanin, etc.
Catalog No. 50-125-1610 Supplier Zymo Research Corporation Supplier No. D6035130
This prepares the columns or plates for the efficient removal of PCR inhibitors such as polyphenolics, humic/fulvic acids, tannins, melanin, etc.