Learn More
Thomas Scientific DirectAmpT Genomic DNA Amplification Kits, Preparation Solution for DirectAmpT Tissue DNA Extraction & Amplification Kit, 100 reactions
SDP
Catalog No. 501459205
Encompass_Preferred
Extract genomic DNA in 15 minutes Tissue, plant or seed Quick isolation and amplification for genotyping Highly specific PCR amplification Eliminates complicated purifications, centrifugation and extractions Denville DirectAmp™ Genomic DNA Amplification Kits contain all reagents for rapid extraction and amplification of genomic DNA from a variety of sources. These kits provide a simple and quick extraction method that eliminates protocol steps that call for liquid nitrogen freezing, enzyme digestions, hazardous organic extractions, column purification, centrifugation or precipitation. Each kit includes Denville Hot Start Blue Mastermix, noted for its highly specific amplification of targeted genomic DNA, without adding loading buffer or tracking dye for gel analysis. Although included as a standard kit component, Hot Start Blue Mastermix is also available separately.
Catalog No. 50-145-9205 Supplier Thomas Scientific Supplier No. C788T93
Extract genomic DNA in 15 minutes Tissue, plant or seed Quick isolation and amplification for genotyping Highly specific PCR amplification Eliminates complicated purifications, centrifugation and extractions Denville DirectAmp™ Genomic DNA Amplification Kits contain all reagents for rapid extraction and amplification of genomic DNA from a variety of sources. These kits provide a simple and quick extraction method that eliminates protocol steps that call for liquid nitrogen freezing, enzyme digestions, hazardous organic extractions, column purification, centrifugation or precipitation. Each kit includes Denville Hot Start Blue Mastermix, noted for its highly specific amplification of targeted genomic DNA, without adding loading buffer or tracking dye for gel analysis. Although included as a standard kit component, Hot Start Blue Mastermix is also available separately.