Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Preparative Chromatography Mix Standard, 1 mL, Reference, Liquid

Catalog No. 50466416 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Preparative Chromatography Mix Standard is a quality control reference material designed to evaluate and benchmark chromatography systems. This standard mix is formulated to confirm the benchmark performance of preparative/purification systems and includes a void marker, as well as neutral, acidic, and basic compounds, making it suitable for use with various column chemistries, dimensions, and system hardware.

  • Detection method: UV, MS
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC, LCMS
  • Quantitative: Yes
  • Shelf life: 12 months
  • Shipping condition: Ambient
  • Volume: 1 mL
  • Type: Reference
  • Application: Small molecule
  • Product type: QC
  • Units per package: 1 pk

Catalog No. 50-466-416 Supplier Waters Corp Supplier No. 186006703
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Preparative Chromatography Mix Standard is a quality control reference material designed to evaluate and benchmark chromatography systems. This standard mix is formulated to confirm the benchmark performance of preparative/purification systems and includes a void marker, as well as neutral, acidic, and basic compounds, making it suitable for use with various column chemistries, dimensions, and system hardware.

  • Detection method: UV, MS
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC, LCMS
  • Quantitative: Yes
  • Shelf life: 12 months
  • Shipping condition: Ambient
  • Volume: 1 mL
  • Type: Reference
  • Application: Small molecule
  • Product type: QC
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.