Learn More
Andwin Scientific PREPARATIVE COMB FOR DNA PLUS
SDP
Catalog No. 50820204
Encompass_Preferred
Preparative comb for DNA Plus;Self-Circulating Standard Horizontal Gel System. Teeth/Comb: 2. Thickness: 1.5 mm. Tooth Width: 5 mm/194 mm. Volume/Well: 35 ul/ 1450 ul.
Catalog No. 50-820-204 Supplier Andwin Scientific Supplier No. 34820500
Preparative comb for DNA Plus;Self-Circulating Standard Horizontal Gel System. Teeth/Comb: 2. Thickness: 1.5 mm. Tooth Width: 5 mm/194 mm. Volume/Well: 35 ul/ 1450 ul.