Learn More
Bio Rad Laboratories PREPARATIVE COMB FOR SUB-CELL
Catalog No. 509012237
Encompass
Preparative Comb; Bio-Rad; For Sub-Cell and Wide Mini-Sub Cell Systems; 1 preparative; 2 reference wells; 1.5mm thick; Fixed height
Catalog No. 50-901-2237 Supplier Bio Rad Laboratories Supplier No. 1704440
Preparative Comb; Bio-Rad; For Sub-Cell and Wide Mini-Sub Cell Systems; 1 preparative; 2 reference wells; 1.5mm thick; Fixed height